Learn More
Waters Corp Q-TOF Premier/SYNAPT Performance Maintenance Kit
Supplier:  Waters Corp 201000187
Encompass_Preferred
Request Bulk Order
The Waters Performance Maintenance Kit for the Q-Tof Premier Mass Spectrometer contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: Source component, ESI probe components, and vacuum pump components. This PM kit is also used for the SYNAPT HDMS and the SYNAPT MS mass spectormeters.
Catalog No. 50-791-580