Learn More
Thermo Scientific™ QMS™ Everolimus Immunoassays, Calibrators and Controls
Obtain results you can count on for immunosuppressant drug monitoring with the Thermo Scientific™ QMS™ Everolimus Immunoassay.
$321.00 - $5931.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Everolimus |
Sample Type | Whole Blood |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
23-278-443
|
Thermo Scientific™
0380000 |
QMS Everolimus Immunoassay. For sale in the U.S.A., Mexico and Brazil. |
Each for $5,931.00
|
|
|||||
23-278-440
|
Thermo Scientific™
0380005 |
QMS Everolimus Calibrator Set. For sale in the U.S.A., Mexico and Brazil. |
Each for $321.00
|
|
|||||
23-278-468
|
Thermo Scientific™
0373878 |
QMS Everolimus Control Set |
Each for $578.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Obtain results you can always count on for the crucial monitoring of everolimus drug levels in kidney and liver transplant patients with the Thermo Scientific™ QMS™ Everolimus Immunoassay. Using our proprietary QMS technology, these liquid, ready-to-use reagents provide excellent accuracy and precision. Applications are available on our compact Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Key Features- Optimal low-end sensitivity with a broad dynamic range
- Excellent correlation with HPLC-MS, the gold standard method
- Precise throughout the standard assay range
Specifications
2°C to 8°C | |
Everolimus | |
Whole Blood |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.