Learn More
Air Science Usa Air Science Purair™ FLOW Vertical Laminar Flow Cabinet Clean Bench, 36" / 900mm Nominal Width, 230V 50HZ
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Air Science Usa FLOW36G
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Purair FLOW Vertical Laminar Flow Cabinet (FLOW-36-G), 36" / 900mm Nominal Width, 230V 50Hz (other voltages available). Purair FLOW Series Vertical Laminar Flow Hoods / Clean Bench are designed to protect the work surface, products and materials from particulate contamination. Room air passes through a HEPA filter then uniformly by vertical laminar flow through the cabinet interior to protect the work from unfiltered air. The airflow is oriented to exhaust airborne particulate introduced by the user. The Purair FLOW series product line employs the Air Science exclusive Multiplex HEPA filtration technology to sustain the contamination-free environment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.