Learn More
Electron Microscopy Sciences PTFE Beaker Cap, 500 ml
SDP
Supplier:  Electron Microscopy Sciences 60948C
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 500 ml
Catalog No. 50-189-8980
Explore available promotions