Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ 3 in. RED MSDS BINDER PACKAGE
Package includes binder only (information center and accessories sold separately)
Supplier: Brady™ BR363R
Description
3 in. RED MSDS PACKAGESpecifications
Binder | |
Binder; Inserts | |
347.98 mm | |
Each | |
Polyethylene | |
English |
Binder | |
3 in. | |
11 in. | |
3.5 in. | |
Yellow on Red | |
Hazard Communication |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction