Learn More
Thermo Scientific Pressure Transducer 1 EA
Supplier: Thermo Scientific 067438
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
pressure transducer with piezoresistive silicon sensor measures positive or negative gauge pressure for monitoring hydraulic systems, gas flow, liquid levels & industrial pressure, withstands 5000 psi & temperatures from -40 to 257°F
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)