Learn More
SCS Premium 3-Layer Vinyl Mat Roll
![SDP](/content/dam/fishersci/glyphs/sdp.png)
This three-layer vinyl mat provides excellent electrical properties.
Supplier: SCS TM301200L3BL
Description
- The top layer is made from a tough, easy-to-clean dissipative vinyl
- The center layer, a highly conductive metal scrim, moves the charge from the surface to ground quickly
- The bottom layer is a durable, non-skid, cushion of dissipative foam
- Compatible with constant monitors
- Meets ANSI/ESD S20.20 worksurface required limits and recommendation of ANSI/ESD S4.1
Specifications
Blue | |
Vinyl | |
0.135 in. | |
100 ft. |
30 in. | |
100 ft. x 30 in. | |
76.2 cm | |
30.48 m |