Learn More
Kamiya Biomedical Prelabumin Calibrator, 6x1mL/PK RX
Supplier: Kamiya Biomedical KAI054C
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
K-ASSAY Prealbumin Calibrator. 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY Prealbumin Calibrator is intended to be used for the calibration of the K-ASSAY Prealbumin immunoturbidimetric assay (Cat. No. KAI-053) for quantifying prealbumin in serum specimens. FOR IN VITRO DIAGNOSTIC USE.