Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Non-Skid Polypropylene Shoe Covers

Catalog No. 19472499
Click to view available options
Non-Skid Polypropylene Shoe Covers; Size: X-Large; Closure Type: Elastic; Material: Polypropylene; Color: Blue

Polypropylene shoe covers feature anti-skid soles for better traction and provides protection against dirt, grime, and particulates.

Polypropylene shoe covers safeguard against dirt, grime, and other unwanted particulates in non-hazardous environments. Anti-skid soles provide enhanced traction to help prevent accidental slips. Breathable, lightweight fabric helps keep feet cool and comfortable over long hours. Typically usage includes food processing plants, pharmaceutical labs, or an environmental cleanup sites.

  • Machine made featuring welded, heat-sealed seams
  • Fits up to a men's size 12 shoe or size 11 boot
  • Light blue color mask stains
  • Anti-skid soles for better traction
  • Recommended applications include food processing, pharmaceuticals, environmental cleanup, moving services, realtors, beauty, and more

Specifications

Size X-Large
Material Polypropylene
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Seam Type Welded
Resistance Dry Particulates
Product Type Controlled Environments Shoe Covers
Show More Show Less
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.