Learn More
Pointe Scientific Amylase-EPS Reagents
Supplier:  Pointe Scientific HA965324
 View more versions of this product
Catalog No. 23-666-106
Add to Cart
This item is not returnable. View return policy
Description
Description
HA965-324 is for Hitachi 911, 912, 914. HA765-162 is for Hitachi 704. HA465-R1 and HA465-R2 are for Hitachi 747.
Specifications
Specifications
CE Marked
Buffer 50mM pH 7.2 ±0.1, NaCl >50mM, MgCl2 10mM, alpha-Glucosidase >2KU/L, EPS-G7 >1mM
Hitachi 911, 912, 914 analyzer
Liquid
Amylase-EPS Reagent Kit
Quantitative kinetic determination of a-amylase activity in Human Serum
12 x 22mL R1, 6 x 210mL R2