Learn More
Monarch Instrument PLS STROBOSCOPE KIT
SDP
Supplier:  Monarch Instrument 6235011
Encompass_Preferred
Each Unit Includes • Strobe • Universal 115/230 Recharger with Interchangeable Wall Plugs • Manual • NIST Calibration • Plastic Latching Carry Case Product Highlights • Diagnostic Inspection and RPM Checks • Accepts Remote Sensors • Tripod Mounting Bushing (1/4-20) • CE Marked, RoHS Compliant • NIST Certificate Included • Compact Pocket Sized Package • Intuitive One Hand Operation • Flash Range 30 - 300,000 FPM/RPM • Display Multiline LCD • Accuracy/Resolution .002% of setting or +/- 1 lsd/ 0.01 FPM
Catalog No. NC0732203