Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Platinum Iridium wire, 1.0mm (0.04in) dia, Thermo Scientific Chemicals
$673.00 - $9415.00
Specifications
Chemical Name or Material | Platinum Iridium wire |
---|---|
Name Note | 1.0mm (0.04 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
MDL Number | MFCD02091683 |
Odor | Odorless |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA42678BQ
|
Thermo Scientific Chemicals
042678BQ |
5 cm |
Each for $673.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA42678BU
|
Thermo Scientific Chemicals
042678BU |
25 cm |
Each for $2,939.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA42678BY
|
Thermo Scientific Chemicals
042678BY |
100 cm |
Each for $9,415.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 90:10 wt% | |
Odorless | |
Wire |
1.0mm (0.04 in.) dia. | |
MFCD02091683 | |
≈0.85g/5cm |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title