Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
New Pig Floor Sign and Marker
![SDP](/content/dam/fishersci/glyphs/sdp.png)
ADA Compliant Floor Signs and Markers are easy to install, stay down, and do not leave residue or damage floors. The low profile signs can be cleaned and will not create issues for heavy foot, cart, wheelchair, or walker traffic.
Supplier: New Pig GMM99001BK
Description
- Helps companies and facilities to stay in compliance with social distancing guidelines
- Creates a safe environment
- Communicates to employees, guests & visitors how to safely navigate facilities
- Withstands foot traffic, floor scrubbers, and disinfectant use
- Will not become slippery when wet
- Can be frequently cleaned with a scrubber, vacuum or broom without affecting performance
- Sticks to most commercial floors including smooth indoor concrete, vct, linoleum, ceramic tile, quarry tile, and laminate flooring
- Recommended for indoor & non-carpeted surfaces only
- Lasts up to three months
Specifications
Polypropylene | |
To Remind and Reinforce Maintaining a 6 ft. Distance While Waiting in Line | |
English | |
Play It Smart Keep 6 ft. Apart |
Self-adhesive | |
Square | |
9 x 9 in. | |
Yellow on Black |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction