Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum Foil Sheets | Pack Size: 100 Foils | 78 x 125 mm | Helps Prevent Sample Evaporation and Contamination | Compatible with Lab Automation Equipment | Enables Cold Storage Down to -80°C | 100 Foils/Pack
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z740334100EA
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Aluminum foil sheets 78 x 125 mm | Pack size: 100 foils | Creates airtight seal on plates and tubes | Helps prevent sample evaporation and contamination | Compatible with lab automation equipment | Enables cold storage down to -80°C | Unit of Measure: 100 foils/pack