Learn More
Bioassay Systems PiBlue™ Phosphate Assay Kit. For sensitive and high-throughput phosphate determination.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems POPB500
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For sensitive and high-throughput phosphate determination. Key Features: Reagent very stable. Due to our innovative formulation, no precipitation of reagent occurs. Therefore no filtration of reagent is needed prior to assays, as is often required with other commercial kits. High sensitivity and wide detection range: detection of as little of 20 pmoles of phosphate and useful range between 0.4 µM and 50 µM phosphate. Fast and convenient: single reagent "mix-and-measure". Compatible with routine laboratory and HTS formats: assays can be performed in tubes, cuvettes or microplates, on spectrophotometers and plate readers. Robust and amenable to HTS: Z factors of 0.7 to 0.9 are observed in 96-well plates. Can be readily automated on HTS liquid handling systems Method: OD620nm (malachite green). Samples: Enzyme reactions with free phosphate releases. Species: All. Procedure: Assay takes 30 min. Kit size: 500 tests. Detection limit: 0.4 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.