Learn More
Bioassay Systems QuantiChrom™ Phytase Assay Kit. A simplest, direct "Add-and-Measure" assay for phytase enzyme activity in agricultural and biological samples.
Supplier: Bioassay Systems DPHT100
A simplest, direct "Add-and-Measure" assay for phytase enzyme activity in agricultural and biological samples. Key Features: Simple and convenient: No complex detection reagents to mix. High sensitivity and wide detection range: Detection range of 0.01 to 20 U/L phytase in a 96-well plate assay. Fast and high-throughput: Homogeneous "mix-and-measure" assay allows for quantification of phytase activity within 60 minutes. Method: OD620nm. Samples: Enzyme extracts, agriculture and biological samples. Species: All (e.g. Wheat, Barley). Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 0.01 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.