Learn More
Kent Scientific PhysioSuite™ Monitor Module Upgrade MouseSTAT™ Pulse Oximeter and Heart Rate Monitor
With mouse sensor.
Supplier: Kent Scientific PS03MODULE
Includes: mouse sensor
Description
- Engineered for mice and rats
- Measures heart rates up to 900 bpm
- Soft touch paw sensors
- Integrates with ventilation and end-tidal CO2 monitoring modules
Specifications
Measures heart rates up to 900 bpm | |
PhysioSuite™ Monitor Module Upgrade | |
Pulse oximeter | |
mouse sensor |