Learn More
SKC Inc Surface contamination detector for phenols, pk/25
Supplier: SKC Inc 7691027
Suitable for OSHA PPE Standard. Fast, easy results in 3 minutes. Convenient and safe. Use on almost any surface — Skin SWYPEs are available. Inexpensive – no instruments or analysis required. Available for a variety of chemicals with ACGIH skin notations. Ideal for dermal exposure reduction programs.