Learn More
Saint Gobain Performance Plastics PHARMED BPT PUMP TUBING
Supplier:  Saint Gobain Performance Plastics AY242019
Encompass_Preferred
Saint-Gobain PharMed BPT Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 1/2 in. 12.7 mm Thickness 0.13 in. 3.2 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
Catalog No. 50-206-9039
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product