Learn More
Saint Gobain Performance Plastics PHARMED BPT PUMP TUBING
Supplier: Saint Gobain Performance Plastics AY242605
Saint-Gobain PharMed BPT Tubing Diameter Inner 0.02 in. 0.5 mm Diameter Outer 0.145 in. 3.7 mm Thickness 0.063 in. 1.6 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.