Learn More
Acute Care Pharmaceuticals Pharma-Chemo Mats™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Acute Care Pharmaceuticals PCMS1611
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pharma-Chemo Mats™ are designed to be your defense against spills and contamination. Use our chemo-prep mats in your cleanroom, barrier isolator, or glove box during HD compounding. With it's highly absorbent top layer and its non-porous backing, the Pharma-Chemo Mat™ protects your surfaces. 15 mats per bag, 2 bags, 30 mats per box.