Learn More
Thermo Scientific™ Phadia™ Cardiolipin Antibodies for Antiphospholipid Syndrome
$240.00 - $480.00
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
14-552-801P
View Documents
Thermo Scientific™
14552801
EliA Cardiolipin IgA Wells; Equivocal: 14 - 20 APL-U/mL; Size: 2x12 EliA Cardiolipin IgA Wells; Equivocal: 14 - 20 APL-U/mL; Size: 2x12
Pack of 24 for $240.00
Add to Cart
14-552-901P
View Documents
Thermo Scientific™
14552901
EliA Cardiolipin IgG Wells; Equivocal: 10 - 40 GPL-U/mL; Size: 4x12 EliA Cardiolipin IgG Wells; Equivocal: 10 - 40 GPL-U/mL; Size: 4x12
Pack of 48 for $480.00
Add to cart
14-553-001P
View Documents
Thermo Scientific™
14553001
EliA Cardiolipin IgM Wells; Equivocal: 10 - 40 MPL-U/mL; Size: 4x12 EliA Cardiolipin IgM Wells; Equivocal: 10 - 40 MPL-U/mL; Size: 4x12
Pack of 48 for $480.00
Add to cart
Description
Description
If present in the patient's specimen, antibodies to cardiolipin bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG (IgM,IgA) antibodies (EliA IgG, IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured.To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA Cardiolipin IgG (IgM, IgA) Wells are coated with bovine cardiolipin antigen
  • The higher the response value, the more specific IgG (IgM, IgA) is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'