Learn More
CTL Scientific Supply Corp PH-FIX STRIPS 2-9,CS/10BX/100
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 9211810
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PH-FIX 2.0-9.0 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 2.0 - 2.5 - 3.0 - 3.5 - 4.0 - 4.5 - 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 (case of 10 boxes of 100 strips each)