Learn More
CTL Scientific Supply Corp PH-FIX ST. 7.5-9.5,CS/10BX/100
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 9216010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PH-FIX 7.5-9.5 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 7.5 - 7.9 - 8.2 - 8.4 - 8.6 - 8.8 - 9.1 - 9.5 (case of 10 boxes of 100 strips each)