Learn More
BioMed Diagnostics A pre-prepared medium with an improved formulation including selective antibiotics for isolation and identification of T. equigenitalis. | Detects: Taylorella equigenitalis. | Available in: 10 pack
Supplier: BioMed Diagnostics 12144001
A pre-prepared medium with an improved formulation including selective antibiotics for isolation and identification of T. equigenitalis and can be used in tandem with Chocolate Eugon. | Detects: Taylorella equigenitalis | Time to Result: 18-24 hours, incubate at 37C | Available in: 10 pack