Learn More
BioMed Diagnostics With 10% horse blood, this pre-prepared medium aids in the culture, isolation and identification of fastidious pathogenic bacteria including Taylorella equigenitalis. | Available in: 10 pack
Supplier: BioMed Diagnostics 12134001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
With 10% horse blood, this pre-prepared medium aids in the culture, isolation and identification of fastidious pathogenic bacteria including Taylorella. | Detects: Taylorella equigenitalis | Time to Result: 18-24 hours, incubate at 37C | Available in: 10 pack