Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Permeabilization Medium (Medium B)
Permeabilization Medium (Medium B)
$110.00 - $1076.00
Specifications
Content And Storage | Room Temperature |
---|---|
Type | Permeabilization Medium (Medium B) |
Reagent Type | Permeabilization Reagent |
Shipping Condition | Room Temperature |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
GAS002S5
|
Invitrogen™
GAS002S5 |
5 mL |
Each for $110.00
|
|
|||||
GAS002S100
|
Invitrogen™
GAS002S100 |
1 Each |
Each for $1,076.00
|
|
|||||
Description
Permeabilization Medium (Medium B) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.The FIX & PERM™ Cell Permeabilization Kit is:
View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
The Permeabilization Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.
How It Works
Cells are incubated for 15 minutes at room temperature with Fixation Medium (Medium A) (Cat. No. GAS001S100) and washed. Cells are then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
Specifications
Room Temperature | |
Permeabilization Medium (Medium B) | |
Permeabilization Reagent | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title