Learn More
Chemglass Life Sciences 100mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Coarse Frit, GL 25 Thread
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG186613
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Similar to CG-1864, but with the addition of a lower standard taper inner joint to allow collection into a vessel having a 24/40 outer joint. COARSE porosity fritted glass resin support. A serrated hose connection is sealed on the inner joint for connection to a vacuum source. The “T”-Bore stopcock has an 8mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8mm at the middle ring.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.