Learn More
Ted Pella Inc PELCO Pro Cyanoacrylate Debonder, 20ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ted Pella Inc 19920
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The PELCO Pro Cyanoacrylate Debonder destabilizes and debonds cured cyanoacrylates. Useful for releasing specimens from holders, cleaning cyanoacrylates from surfaces and removing cured cyanoacylates from fingers. Works on all brands of cyanoacrylate glue (Loctite, Henkel, Permabond, 3M™, PELCO, Aron Alpha, etc.). The debonder is also a solvent, please check before using on plastic surfaces / samples and avoid repeated skin contact. Supplied in 20ml bottle with dispenser tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.