Learn More
Ted Pella Inc PELCO High Performance Silver Paste, 50g
Supplier: Ted Pella Inc 16047
Silver flakes in an inorganic silicate aqueous solution, specially formulated adhesive for demanding bonding applications.