Learn More
Electron Microscopy Sciences CoverWell™ Assortment Pack 5 Ea Of A,B,C,D & 10 Of E
Supplier: Electron Microscopy Sciences 7033405
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Flexible, press-to-seal gasket and coverslip enclosures which can be quickly assembled and disassembled for the specialized challenges of hybridizing and imaging thick and free- floating specimens.
All steps from permeabilizing to image analysis are made easy and trouble free with chambers you assembles from modular components.
For neural explants, plant, or animal specimens of any texture and thickness.
Build up to three chambers on one microscope slide. Ideally suited for overnight hybridizations and agitated washing steps. The screen is made from polyester and is 1.6 mm square.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.