Learn More
Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
Supplier:  Thermo Scientific™ P9DS1
 View more versions of this product
Catalog No. 09-518-157C
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Explore available promotions
Description
Description
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel
Features:
  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P9DS Complete System plus Caster
Dual-Gel Vertical Electrophoresis Complete System
9.8 in.
5.1 in.
7.7 in.
Owl
550 mL
14 to 16 cm
16 cm
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories