Learn More
Chemglass Life Sciences 1.7 Cu. Ft. Oven, Vacuum, 120V
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CLS3450017
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
1.7 Cu. Ft. Oven, Vacuum, 120V. The Vacuum ovens are durably constructed with seamless welds & strong bracing. All of the vacuum ovens are built with a stainless steel chamber for exceptional durability. Ovens incorporate solid brass vacuum valves w/ PTFE seats to prevent leaks. The double plenum design exceeds CE safety requirements resulting in a cool outer surface. The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. A selection of gaskets (for specific applications) & a small footprint increase the versatility of these ovens. The unique cross-flow ventilation forces inert gas to fill the entire chamber. Users can choose from 3/8 inch orifices or a KF25 fitting to withstand heavy use, minimize draw-down time and achieve low vacuum levels. We offer a full line of gaskets to suit applications involving high temperatures, acids, or solvents. Ovens have a 24 month limited warranty
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.