Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ OSHA Caution Fold-Ups™ Floor Sign: SLIPPERY FLOOR
Fold-Ups™ Floor Signs quickly and easily identify temporary hazardous conditions or display important messages.
Supplier: Accuform™ MF105
Description
- Ideal for multiple uses and rough handling
- Self-supporting and constructed of high-impact plastic
- Cut-out handles for ease of carrying
- Fold flat for transport and storage
- Both sides are printed with the same message
Specifications
OSHA 29 CFR 1910.145, ANSI Z535 | |
Polystyrene | |
Non-adhesive | |
Rectangular | |
20 x 12 in. | |
Floor Safety Sign |
Non-glare | |
1 Sign | |
Floor-standing | |
English | |
CAUTION SLIPPERY FLOOR | |
Black, Red, Yellow |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction