Learn More
Ortho-Clinical Diagnostics™ A2 Cell
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For detection and identification of blood group antibodies
Supplier: Ortho-Clinical Diagnostics™ 719310
This item is not returnable.
View return policy
Description
- Available in 3% suspension
- Used as additional test in reverse group testing
Specifications
Ortho™ A2 Reagent Red Cells | |
1 x10 mL |