Learn More
Perkin Elmer US LLC Organics Sample Introduction Kit for Avio 200/500
Supplier: Perkin Elmer US LLC N0790610
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This kit includes: 3-Slot Hybrid XLT Torch, Qty. 2 (N0790249) 1.2 mm I.D. Alumina Injector, Qty. 2 (N0791181) Baffled Cyclonic Spray Chamber, Qty. 1 (N0791352) Low-Flow GemCone Nebulizer, Qty. 1 (N0770358) Injector Adapter Kit, Qty. 1 (N0790141)