Learn More
Chemglass Life Sciences 40-Position Block for 12 x 75mm Round Bottom Tubes
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences OP660034
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
40-Position Block for 12 x 75mm Round Bottom Tubes. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.