Learn More
Ampco Safety Tools Open End Wrench (Imperial), < 2 in.
SDP
AMPCO™ Open End Wrench performs in high-risk and demanding environments.
Supplier:  Ampco Safety Tools 0256
 View more versions of this product
Catalog No. 19-173-231
Explore available promotions
Add to Cart
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Description
Description
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Specifications
Open End
Aluminum Bronze
Factory Mutual (FM) Approved
13/32 in.
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.