Learn More
Showa Denko America Inc OHpak LB-803
Supplier: Showa Denko America Inc F6429201
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Polyhydroxymethacrylate packed columns for aqueous SEC (GFC) analysis, low column bleeding allows its use with light scattering detectors, the eluent can be replaced with DMF enabling the analysis of polar polymers, fulfills USP L38 and L39 requirements, dimensions: 8.0 mm ID x 300 mm L, target MW range: 1,000 − 100,000 Daltons