Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ohio Valley Specialty Chemical Specialty Chemical Snoop™ Liquid Leak Detector
Used for locating leaks in air and carrier gas lines
Supplier: Ohio Valley Specialty Chemical 21011
Description
- 236mL (8 oz.) squeeze bottle with 30.5cm (12 in.) snooper tube
- Nontoxic, nonflammable and noncorrosive
- Temperature range: -2 to +93°C (27 to 200°F)
- For external surface use only
Specifications
Leak Detector | |
Locating leaks in air and carrier gas lines |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction