Learn More
LGC Biosearch Technologies NxGen T4 DNA Ligase (High Concentration), 1500 U @ 10 U/LA workhorse, high quality DNA ligase for joining blunt or cohesive ends of dsDNA or dsRNA.
Supplier: LGC Biosearch Technologies 302431
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
NxGen T4 DNA Ligase (High Concentration), 1500 U @ 10 U/LReliably join double-stranded nucleic acid chains.The low concentration ligase includes NxGen T4 DNA Ligase at 2 U/L and 10X T4 DNA ligase Buffer. The high concentration ligase includes NxGen T4 DNA Ligase at 10 U/L and both 2X Rapid Ligation Buffer and 10X T4 DNA ligase Buffer.
This item is not returnable.
View return policy