Learn More
United States Biological Corporation Nonidet-P40 substitute, 9036-19-5, 500mL
Supplier: United States Biological Corporation N3500500ML
Reagent Grade, Molecular Weight 603, Nonidet-P40 is an anhydrous liquid nonionic surface- active agent produced by the reaction of octyl phenol with 8.5-9.5 moles of ethylene oxide. Nonidet-P40 (NP-40) is a nonionic surfactant used in the isolation of membrane complexes. This product has been reformulated to be eco-friendly. The only observable differences are that the viscosity and handling characteristics are somewhat modified. Due to its nonionic structure, this product is compatible with anionic surfactants and is stable in the presence of acids, bases, and salts. It should not be mixed with concentrated oxidizing or reducing agents since the mixture of these compounds with organic compounds could form a potentially explosive mixture. Nonidet-P40 is an effective emulsifier for solvents such as xylene.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.