Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ TBE Gels, 6%

Novex™ TBE Gels 6% are designed to run on the XCell SureLock™ Mini-Cell and provide high-resolution analysis of restriction digests and PCR products.

$214.00 - $234.00

Specifications

Content And Storage • Catalog number refers to a single box of 10 gels

Store at 4°C.
For Use With (Equipment) XCell SureLock Mini-Cell
Gel Percentage 6%
Gel Type TBE
Product Type TBE Gel
View More Specs

Products 3
Catalog Number Mfr. No. Description Wells Price Quantity & Availability  
Catalog Number Mfr. No. Description Wells Price Quantity & Availability  
EC6261BOX
Encompass_Preferred
View Documents
Invitrogen™
EC6261BOX
Novex TBE Gels, 6%, 1-well 1-well
Case for $214.00
Add to Cart
 
EC6265BOX
Encompass_Preferred
View Documents
Invitrogen™
EC6265BOX
Novex TBE Gels, 6%, 10-well 10-well
Case for $219.00
Add to Cart
 
EC62655BOX
Encompass_Preferred
View Documents
Invitrogen™
EC62655BOX
Novex TBE Gels, 6%, 15-well 15-well
Case for $234.00
Add to Cart
 
Description

Description

Novex™ TBE Gels 6% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 65–250 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium­ bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results

Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
6%
TBE Gel
10 Gels/Box
Wet Ice
XCell SureLock Mini-Cell
TBE
65 to 250 bp
Double-stranded DNA (dsDNA)
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.