Learn More
Nextteq™ Gastec™ Airflow Indicator Kit
SDP
Contains everything needed to perform multiple respirator fit tests without the cost of extra equipment
Supplier:  Nextteq™ NX9500/DEL
Catalog No. 19-024856
Add to Cart
Includes: Kit Contents: Six smoke tubes, aspirator bulb pump, tube tip breaker pins, tube protector, manual, carrying case
Description
Description
A quick, simple and economical kit, avoiding the need for chamber construction, nebulizers, electric pumps, mixing jars, batteries, special test masks, mask probes or testing hoods.
  • Simple and effective method for tracing and determining the source, direction, and dispersion of air currents
  • Highly visible white smoke provides an immediate and obvious cue to the directional pattern of air currents.
  • Intrinsically safe and ready to use, anywhere and anytime
  • No pump calibration required, no flow rate to set
  • Kit simulates real working conditions when used for fit testing
  • Provides up to 50 tests
Compliance
Designed to meet and be used in accordance with OSHA 29 CFR 1910.134
Specifications
Specifications
Airflow Indicator Kit
Kit Contents: Six smoke tubes, aspirator bulb pump, tube tip breaker pins, tube protector, manual, carrying case
Product Suggestions
Product Suggestions