Learn More
Azzota Corp Neutral Density Filter Spectrophotometer Calibration Tools Use For Testing Spectrophotometer Photometric Accuracy Check Multi-Wavelength At 437Nm, 546Nm And 640Nm Filter Size 10 X 10 X 45 mm
Supplier: Azzota Corp Q801
Neutral Density Filter Spectrophotometer calibration tools Use for testing Spectrophotometer Photometric Accuracy Check Multi-Wavelength at 437nm, 546nm and 640nm Filter size 10 x 10 x 45 mm
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'