Learn More
Saint Gobain Performance Plastics ND-100-65 1/8 x 3/16 50FT/PK
Supplier: Saint Gobain Performance Plastics ADF00006
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon ND 100-65 Medical Tubing Diameter Inner 1/8 in. 3.2 mm Diameter Outer 3/16 in. 4.8 mm Overall Length 50 ft. 15.2 m Thickness 1/32 in. Non-DEHP Must order minimum quantity of 2.