Learn More
Biotium NBD C6-ceramide
Supplier: Biotium 60030
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
NBD C6-ceramide (6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosine) has been used for following sphingolipid metabolism in cells, and as a selective stain for the Golgi apparatus in live and fixed cells.Properties: Ex/Em(MeOH) = 466/536 nm; Dark solid soluble in chloroform or DMSO; Store at -20°C and protect from light; C30H49N5O6; MW: 575.74; [94885-02-6].
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.