Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Confined Space Keep Out Signs
Specifications
Certifications/Compliance | OSHA 29 CFR 1910.145 |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
For Use With (Application) | Danger signs should be used when an immediate hazard exists which pose a high probability of severe injury |
Displayed Text | DANGER Confined Space Keep Out |
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity & Availability | ||||
18-999-704
|
National Marker™
D372R |
7 x 10 in. | Plastic | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
18-999-706
|
National Marker™
D372P |
7 x 10 in. | Vinyl | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
18-999-705
|
National Marker™
D372RB |
10 x 14 in. | Plastic | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
Specifications
OSHA 29 CFR 1910.145 | |
Surface Mount | |
DANGER Confined Space Keep Out |
1 Ea. | |
Danger signs should be used when an immediate hazard exists which pose a high probability of severe injury | |
Safety Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title