Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard: No Food or Drink to be Stored Sign

Warns employees to not use refrigerator for food or drink

Supplier:  National Marker™ S71R

Catalog No. 18-999-766

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.


Description

Description

  • Material: Rigid plastic
  • Size: 17.7 × 17.7cm (7 × 7 in.)
  • Color: Orange sign with black text and graphics

Specifications

Specifications

Food and Drink Prohibited Sign
Plastic
7 in.
BIOHAZARD NO FOOD OR DRINK TO BE STORED IN THIS REFRIGERATOR
1 Ea.
7 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit