Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: No Eating, Drinking, Smoking Signs
Specifications
Certifications/Compliance | OSHA |
---|---|
Quantity | 1 Ea. |
Height (English) | 7 in. |
Width (English) | 17 in. |
Displayed Text | BIOHAZARD EATING, DRINKING, SMOKING, APPLYING COSMETICS AND HANDLING CONTACT LENSES ARE PROHIBITED IN THIS AREA |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
18-999-754
|
National Marker™
SA186R |
Plastic | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
18-999-755
|
National Marker™
SA186P |
Vinyl |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Color: Orange sign with black text
- Material: Rigid plastic or pressure-sensitive vinyl
- Size: 17.7 × 43.2cm (7 × 17 in.)
Specifications
OSHA | |
7 in. | |
BIOHAZARD EATING, DRINKING, SMOKING, APPLYING COSMETICS AND HANDLING CONTACT LENSES ARE PROHIBITED IN THIS AREA |
1 Ea. | |
17 in. | |
Safety Sign |
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'